missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalimide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.
Solubility
Slightly soluble in water. Soluble in aqueous alkali hydroxides
Notes
Store at room temperature. Ensure good ventilation.
Phthalimide is a reagent used to transform allyl- and alkyl halides into protected primary amines. . It is used as a precursor to other organic compounds as a masked source of ammonia.
Solubility
Slightly soluble in water. Soluble in aqueous alkali hydroxides
Notes
Store at room temperature. Ensure good ventilation.
Specifications
Specifications
Melting Point | 232°C to 236°C |
Density | 1.21 |
pH | 3.8 |
Boiling Point | 366°C |
Flash Point | 165°C (329°F) |
Odor | Odorless |
Quantity | 500 g |
Beilstein | 118522 |
Merck Index | 14,7373 |
Solubility Information | Slightly soluble in water; Soluble in aqueous alkali hydroxides |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'