missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalocyanine, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Pthalocyanine is a common macrocylic blue-green dye. It is also used as a suitable laser dye metal. Phthalocyanines have long been examined as catalysts for redox reactions. Areas of interest are the oxygen reduction reaction and the sweetening of gas streams by removal of hydrogen sulfide. Phthalocyanine compounds have been investigated as donor materials in molecular electronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Odor | Odorless |
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 514.52 |
Physical Form | Crystalline |
Chemical Name or Material | Phthalocyanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'