Learn More
Phthaloyl chloride, 94%, Thermo Scientific Chemicals
£36.50 - £123.00
Chemical Identifiers
CAS | 88-95-9 |
---|---|
Molecular Formula | C8H4Cl2O2 |
Molecular Weight (g/mol) | 203.018 |
MDL Number | MFCD00000666 |
InChI Key | FYXKZNLBZKRYSS-UHFFFAOYSA-N |
Synonym | phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride |
PubChem CID | 6955 |
IUPAC Name | benzene-1,2-dicarbonyl chloride |
SMILES | C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11332696
|
Thermo Scientific Alfa Aesar
L08753.22 |
100 g |
£36.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342696
|
Thermo Scientific Alfa Aesar
L08753.36 |
500 g |
£123.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phthaloyl chloride is used for the preparation of volatile acid chlorides from the acids. It acts as a trapping agent for the nitrite ion in the fluorodenitration of aromatic substrates with potassium fluoride. It serves as an additive in polymers and fibers to improve the color stability. It is used as an ingredient in ceramic mold binders, as a stabilizer for polymers and in reverse osmosis membranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthaloyl chloride is used for the preparation of volatile acid chlorides from the acids. It acts as a trapping agent for the nitrite ion in the fluorodenitration of aromatic substrates with potassium fluoride. It serves as an additive in polymers and fibers to improve the color stability. It is used as an ingredient in ceramic mold binders, as a stabilizer for polymers and in reverse osmosis membranes.
Solubility
Miscible with ether.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, alcohols oxidizing agents and strong bases.
Chemical Identifiers
88-95-9 | |
203.018 | |
FYXKZNLBZKRYSS-UHFFFAOYSA-N | |
6955 | |
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl |
C8H4Cl2O2 | |
MFCD00000666 | |
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride | |
benzene-1,2-dicarbonyl chloride |
Specifications
88-95-9 | |
1.409 | |
151°C (304°F) | |
1.568 | |
UN3265 | |
Moisture sensitive | |
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride | |
FYXKZNLBZKRYSS-UHFFFAOYSA-N | |
benzene-1,2-dicarbonyl chloride | |
6955 | |
94% |
12°C | |
276°C | |
C8H4Cl2O2 | |
MFCD00000666 | |
608200 | |
14,7375 | |
Miscible with ether. | |
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl | |
203.018 | |
203.03 | |
Phthaloyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H312-H314-H317-H334-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 201-869-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only