Learn More
Thermo Scientific™ Phusion Buffers
Shop All Thermo Scientific ProductsDescription
Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases: 5X Phusion HF Buffer and 5X Phusion GC Buffer. The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.
5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Includes:
Detergent-free 5x Phusion HF Buffer, 50 mM MgCl2 solution, 100% DMSO.
Specifications
Specifications
| Description | Phusion™ HF Buffer Pack |
| For Use With (Application) | Standard PCR |
| Quantity | 3mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.