missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Buffers

Product Code. p-4527011 Shop All Thermo Scientific Products
Click to view available options
Description:
Phusion™ HF Buffer Pack
Quantity:
3mL
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 11508475

Brand: Thermo Scientific™ F518S

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Choose between two buffer packs to optimize high-fidelity PCR or difficult or long template amplification using Thermo ScientificTM PhusionTM High-Fidelity DNA Polymerases.

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases: 5X Phusion HF Buffer and 5X Phusion GC Buffer. The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Detergent-free HF Buffer for Phusion DNA Polymerases Developed for DHPLC and microarray applications

Includes:

Detergent-free 5x Phusion HF Buffer, 50 mM MgCl2 solution, 100% DMSO.

TRUSTED_SUSTAINABILITY

Specifications

Description Phusion™ HF Buffer Pack
For Use With (Application) Standard PCR
Quantity 3mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.