Learn More
Thermo Scientific™ Phusion™ HF and GC Buffer Packs
5X Phusion Green buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel.
Brand: Thermo Scientific™ F520L
22.50 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.
5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.
Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:
- 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
- 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))
Specifications
• 5X Phusion HF Reaction Buffer, Detergent-free (4 x 1.5 mL) • 50 mM MgCl2 (4 x 1.5 mL) • DMSO (0.5 mL) Store at –20°C. |
|
Phusion High-Fidelity DNA Polymerase | |
4 x 1.5 mL | |
Standard PCR, High-fidelity PCR | |
4 x 1.5 mL |
Detergent-free Phusion™ HF Buffer Pack | |
Detergent-free FF Buffer Pack | |
Wet or Dry Ice | |
DNA | |
Colorless |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.