missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ Plus DNA Polymerase

Product Code. 17218570 Shop All Thermo Scientific Products
Click to view available options
No. of Reactions:
100 Reactions
2000 Reactions
500 Reactions
Unit Size:
100 reactions
500 reactions
Pack of 4
This item is not returnable. View return policy

Product Code. 17218570

Brand: Thermo Scientific™ F630S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing.

Thermo Scientific Phusion Plus DNA Polymerase is a hot-start, high-fidelity DNA polymerase that brings together protein fusion technology and universal primer annealing. It is a Pyrococcus-like proofreading DNA polymerase fused to a processivity-enhancing domain, enabling Phusion Plus DNA Polymerase to generate PCR sequences with high accuracy, sensitivity, and inhibitor tolerance.

Features include:

  • >100X higher sequence accuracy than Taq polymerase
  • Universal annealing temperature of 60°C in PCR
  • Hot-start mechanism reduces non-specific amplification and primer degradation
  • Automation enabled due to 24-hour stability of assembled reactions
  • High inhibitor tolerance for work with DNA of suboptimal purity

The novel Phusion Plus reaction buffer includes proprietary additives that stabilize primer/template duplexes during the annealing step at 60°C. This unique buffer formulation increases PCR specificity without the need to optimize annealing temperature for each primer pair, helping save time and avoid mistakes in PCR runs. Furthermore, with Phusion Plus DNA Polymerase and its universal annealing feature, different PCR reactions can be cycled together using the same protocol.

Phusion Plus DNA Polymerase is inactive at ambient temperatures due to its hot-start mechanism mediated by Affibody molecules, allowing setup and storage of assembled PCR reactions at room temperature. Enzyme activity is fully restored after the initial denaturation step.

Applications

  • High-fidelity PCR
  • Cloning & mutagenesis
  • Template generation for sequencing
  • GC-rich PCR
  • Long-range PCR (up to 20 kb)
  • High-throughput PCR

Notes

  • To simplify reactrion setup, Phusion Plus DNA Polymerase is also available in ready-to-use master mix formats.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Phusion Plus DNA Polymerase (50 μL)
• 5X Phusion Plus Buffer (1.25 mL)
• 5X Phusion GC Enhancer (1.25 mL)

Store at –15°C to –25°C.
GC-Rich PCR Performance High
Polymerase Phusion Plus DNA Polymerase
Reaction Speed 15 to 30 sec./kb
Quantity 100 x 50 μL reactions
Shipping Condition Room Temp or Wet Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) >100X
Hot Start Built-In Hot Start
No. of Reactions 100 Reactions
Overhang Blunt
Reaction Format Separate components
Size (Final Product) 20 kb or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.