missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHYHIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16984-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody recognizes both PHYHIP (35-38 kDa) and PHYHIPL (43-45 kDa).
Its interaction with PHYH suggests a role in the development of the central system.Specifications
PHYHIP | |
Polyclonal | |
Unconjugated | |
PHYHIP | |
DYRK1AP3, KIAA0273, PAHX AP, PAHX AP1, PAHXAP1, PHYHIP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
105653, 290356, 9796 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q568Z9, Q8K0S0, Q92561 | |
Phyhip | |
PHYHIP Fusion Protein Ag10575 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |