missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Phytone™ Peptone
Click to view available options
Quantity:
454 g
10 kg
Unit Size:
10kg
454g
Description
- An animal origin-free (AOF) enzymatic digest of soy
- A nutritious and excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
- Customers have successfully used this supplement in CHO-based production of monoclonal antibodies for oncological and rheumatological therapeutics
- Also used with many different microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines
Applications of Phytone Peptone include:
- It is an excellent plant peptone for the cultivation of fungi and fastidious types of bacteria, such as members of the Clostridium and Neisseria genera
- It has been used in mammalian cell culture applications due to its high carbohydrate content
Specifications
Specifications
| Certifications/Compliance | LUO |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Format | Bottle |
| For Use With (Application) | Production of monoclonal antibodies for oncological and rheumatological therapeutics, microbial cell types to produce animal health vaccines for swine, such as Clostridial vaccines |
| Product Line | Phytone |
| Quantity | 454 g |
| Source | Animal origin-free, soy |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction