missing translation for 'onlineSavingsMsg'
Learn More

PI 828, Tocris Bioscience™

Product Code. 18055882
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

PI 3-kinase inhibitor, more potent than LY 294002 (Cat. No. 1130)

PI 3-Kinase inhibitor (IC50 values are 0.098, 0.183, 0.227 and 1.967 μM for p110β, p110α, p110δ and p110γ respectively) that displays higher potency than LY 294002 (Cat No. 1130). Can be immobilized onto a solid phase.

Chemical Identifiers

CAS 942289-87-4
Molecular Formula C19H18N2O3
Molecular Weight (g/mol) 322.364
InChI Key WUKMIBOGGXMBAC-UHFFFAOYSA-N
Synonym 8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one
PubChem CID 25181195
IUPAC Name 8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one
SMILES C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N

Specifications

Quantity 1 mg
Formula Weight 322.36
Percent Purity >99%
Chemical Name or Material PI 828
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'