Learn More
PI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535878
Code : NEW
Additional Details : Weight : 0.00910kg
Description
Recommended positive controls: human PI3-transfected 293T. Predicted reactivity: Rhesus Monkey (94%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PI3 Polyclonal antibody specifically detects PI3 in Human samples. It is validated for Western Blot.Specifications
PI3 | |
Polyclonal | |
Unconjugated | |
PI3 | |
cementoin; Elafin; elastase-specific inhibitor; ESI; peptidase inhibitor 3; peptidase inhibitor 3, skin-derived; PI3; PI-3; pre-elafin; protease inhibitor 3, skin-derived (SKALP); protease inhibitor WAP3; SKALP; skin-derived antileukoproteinase; trappin-2; WAP four-disulfide core domain 14; WAP four-disulfide core domain protein 14; WAP3; WFDC14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5266 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P19957 | |
PI3 | |
Full length human Elafin Recombinant protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |