missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PichiaPink™ Media Kit

PichiaPink™ Media Kit

Brand:  Invitrogen™ A11156

557.60 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10116593

  • £656.00 / Each
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The PichiaPink™ Media Kit is a set of convenient prepackaged media pouches for use with the PichiaPink™ Yeast Expression System. The PichiaPink™ Media Kit contains media needed to grow Pichia pastoris strains from frozen stocks to cultures ready for transformation and selection. The PichiaPink™ Media Kit is an easy way get started using Pichia pastoris for protein production. Use the PichiaPink™ Media Kit to refill your PichiaPink™ Yeast Expression System Kits.

The PichiaPink™ Media Kit comes with prepackaged pouches for the following solutions. Each pouch makes 1 liter of solution. Media pouches should be kept dry and stored at room temperature.

Why Choose the PichiaPink™ Yeast Expression System?
The PichiaPink™ Yeast Expression System is based on the yeast Pichia pastoris. Advantages of Pichia pastoris include rapid growth, well-defined genetic background, simple media formulation, and easy handling. For over 30 years, Pichia pastoris has been used by labs around the world for producing hundreds of different proteins from many species including human (Ref 1, 2). The PichiaPink™ Yeast Expression System allows convenient and cost-effective protein production from small to large scales.

For information on obtaining a commercial-use license for the PichiaPink™Yeast Expression System, please inquire at outlicensing@lifetech.com.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Related Links
  • Learn more about the PichiaPink™ Yeast Expression System.
  • Learn more about our other protein expression systems.

    References

    1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
    2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains pouches sufficient to make 4 basic media:

    2 pouches PAD agar, 2 pouches YPDS, 2 pouches YPD, 2 pouches YPD agar, 1 pouch Dextrose


    Store at room temperature

    Kit
    Autoclave
    Liquid Medium, Agar Plates
    1 Kit
    PichiaPink™
    Powder
    PAD, YP, YPS
    P. pastoris
    A11150, A11151, A11152, A11153
    Room Temperature
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.