missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PichiaPink™ Secreted Vector Kit
PichiaPink™ Secreted Vector Kit
Brand: Invitrogen™ A11153
Code : 50
Additional Details : Weight : 0.09000kg
Description
The PichiaPink™ Secreted Vector Kit contains the pPINKα-HC vector for use with the PichiaPink™ Yeast Expression System. The pPINKα-HC vector is built around the ADE2 gene for complementing the complementing the ade2 deletion in the PichiaPink™ strains. The pPINKα-HC vector has the α-mating factor secretion signal sequence so that all proteins are secreted. You can order the PichiaPink™ Secreted Vector Kit to refill your PichiaPink™ Yeast Expression System Kit.The PichiaPink™ Secreted Vector Kit comes with:
Why Choose the PichiaPink™ Yeast Expression System?
The PichiaPink™ Yeast Expression System is based on the yeast Pichia pastoris. Advantages of Pichia pastoris include rapid growth, well-defined genetic background, simple media formulation, and easy handling. For over 30 years, Pichia pastoris has been used by labs around the world for producing hundreds of different proteins from many species including human (Ref 1, 2). The PichiaPink™ Yeast Expression System allows convenient and cost-effective protein production from small to large scales.
For information on obtaining a commercial-use license for the PichiaPink™Yeast Expression System, please inquire at outlicensing@lifetech.com.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Related Links
References
1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
Specifications
Liquid | |
AOX1 | |
PichiaPink™ Secreted Protein Vector kit contains 20 μg pPinkα-HC (the high-copy number plasmid with the S. cerevisiae Alpha mating factor presequence) with sequencing primers 5' α-factor and 3' CYC1. Store at -20°C. | |
No Cleavage Site | |
Room Temperature | |
Restriction Enzyme/MCS |
Methanol | |
Vector Kit | |
Ampicillin (AmpR) | |
pUC Origin | |
Untagged | |
pPink |
For Research Use Only. Not for use in diagnostic procedures.