Learn More
PICK1 Mouse anti-Human, Unconjugated, Clone: 2E11A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67099-1-IG-150UL
This item is not returnable.
View return policy
Description
The PICK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PICK1. This antibody recognizes human, mouse, rat, pig antigen. The PICK1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
PICK1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, ELISA | |
2E11A4 | |
Unconjugated | |
150 μL | |
9463 | |
IgG2a |