missing translation for 'onlineSavingsMsg'
Learn More
Picric Acid R1 Solution, EU Pharmacopoeia Grade, Fisher Chemical™ Product Code.: 12950494

Picric Acid R1 Solution, EU Pharmacopoeia Grade, Fisher Chemical™

Product Code. 12950494
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 12950494

Brand: Fisher Chemical RP 158 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 100 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1094029 Certificate TypeCertificate of Analysis Date16/03/2025 Catalog NumberRP 158 100
Lot Number1079398 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberRP 158 100
Lot Number1078833 Certificate TypeCertificate of Analysis Date05/03/2025 Catalog NumberRP 158 100
Lot Number1065836 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 158 100
Lot Number1004154 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 158 100
Product Identifier
  • Picric acid solution R1. European Pharmacopoeia Ref: 1065802 (RP 158)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Picric Acid R1 Solution, EU Pharmacopoeia Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.