Learn More
Sartorius PICUS™ 12-Channel Electronic Pipet
GREENER_CHOICEBIOPHARMA
Product Code. p-4893712 Shop All Sartorius Products
missing translation for 'orderingAttributeHoverText'
Color:
Gray
Orange
Purple
Yellow
Volume (Metric):
0.2 to 10 μL
10 to 300 μL
5 to 120 μL
50 to 1200 μL
Increments:
0.02μL
0.1μL
0.2μL
1μL
Precision:
1.00 to 0.20
1.50 to 0.20
3.00 to 0.50
missing translation for 'unitSize'
Each
Product Code. 13233029
missing translation for 'mfr': Sartorius 735441
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduces risk of repetitive strain injury. Sartorius PICUS™ 12-Channel Electronic Pipet ensures the most accurate, precise and reliable results due to advanced DC-motor concept and built-in error control.
Light and small electronic pipet ensures an optimal working posture and offers comfortable pipeting. New ergonomic design protects from RSI (repetitive strain injury) and eases workload in long pipetting series. Provides more reliable results with a new generation technology and error control system.
  • Helps prevent repetitive strain injury (RSI) due to lightweight, ergonomic design and electronic tip ejection
  • Ensures the most accurate, precise and reliable results due to advanced DC-motor concept and built-in error control
  • Saves time with extremely fast one-hand adjustment, a unique built-in microplate tracker and easy programmability
  • Adjustment wheel for scrolling menus, adjusting volume and controlling manual pipetting
  • Variety of pipetting modes in user friendly interface including memory places for favorite programs
  • Unique Tracker in conjunction with pipetting, reverse pipetting and multi dispensing modes to help microplate work
  • Adjustment possible for 1-point, 2-points and 3-points
  • Easy maintenance of lower parts with GLP information integrated in software for service and calibration
  • Safe-Cone filters in all models >10 μL to prevent contamination
  • Autoclavable lower parts in all models (excluding 1200 μL model)
  • Compatible with electronic pipet charging systems, charging stand for one pipet and charging carousel for four pipets
  • Direct charging option with micro USB cable
  • Delivered without USB AC adapter (available separately)
TRUSTED_SUSTAINABILITY
Specifications
Accuracy 0.5%, 0.7%, 2%
Compatible Tips 200, 350μL Tip
Autoclavable Autoclavable
Color Yellow
Number of Channels 12
Product Line PICUS
Programmability Yes
Type Electronic Pipette
Volume (Metric) 5 to 120 μL
Finger Style Ergonomic
Increments 0.1μL
Precision 1.50 to 0.20
Quantity Each
Warranty 2 Years