missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ 2 Electronic 12-Channel Pipette
GREENER_CHOICEBIOPHARMA

Product Code. 17898549 Shop All Sartorius Products
Click to view available options
Volume (Metric):
10 μL
120 μL
300 μL
1200 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17898549

Brand: Sartorius LH747461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Picus™ 2 Electronic 12-Channel Pipette offers reliable, ergonomic pipetting for all users. It has customizable programs, safety features, and connects to the Sartorius app for automatic adjustments and improved workflow, enhancing lab productivity.

  • Picus™ 2 electronic 12-channel pipettes offer reliable performance, while also being kind to your hand. The unmatched ergonomic design not only reduces strain, but also helps to ensure reproducible pipetting results.
  • Easily accessible pipetting modes and customizable programs for popular applications save you time in the lab.
  • Save time: Picus™ 2 Electronic Pipette saves you time in the lab with a wide selection of pipetting modes and customizable programs to meet your every need. The most frequently used modes with specified settings can be saved on your device for quick and easy access. By connecting Picus™ 2 to your mobile device, you can use the Sartorius pipetting mobile app to smoothly run sample preparation workflows and adjust the pipette setting automatically, taking your productivity to the next level.
  • Reliable results: Picus™ 2 Electronic 12-channel pipettes offer reliable performance while being kind to your hand. Being extremely light and compact for 12-channel pipettes the users don't feel the fatigue as easily, leading more accurate results. The build in plate tracker helps the user to keep track on progress with 96- and 384-well plates.
  • Safety features: Safety features like password protection and calibration reminders support compliance requirements in regulated labs. Pipettes can also be locked from use, in when awaiting service or cleaning.
  • The Picus™ 2 12-channel pipettes can be decontaminated with disinfectants, UV, or autoclaving (excluding 1,200L models).
  • Reliable and repeatable pipetting results
  • A wide selection of pipetting modes
  • Customizable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Sartorius Optifit Tips, SafetySpace Tips, Flexibulk Tips
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Optifit Tip (10μL), SafetySpace Filter Tip
Grip Type None
Material Polycarbonate ABS
Number of Channels 12
Programmability Yes
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 300 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.