missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Picus™ 2 Electronic 8-Channel Pipette
GREENER_CHOICEBIOPHARMA

Product Code. 17898469 Shop All Sartorius Products
Click to view available options
Volume (Metric):
10 μL
120 μL
300 μL
1200 μL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17898469

Brand: Sartorius LH747361

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Picus™ 2 Electronic 8-Channel Pipette offers reliable, ergonomic pipetting for all users. It has customizable programs, safety features, and connects to the Sartorius app for automatic adjustments and improved workflow, enhancing lab productivity.

  • Ensures reliable and repeatable pipetting results and features an unbeatable ergonomic design that is kind to your hand
  • Picus™ 2 is suited to anyone working in the lab, from a graduate student to an experienced laboratory professional. It is as intuitive to use as a mechanical pipette, but also offers more advanced options for experienced users.
  • Speed up your work: Speed up work by choosing from one of the multiple pipetting modes, which are specifically tailored to ease repetitive work.
  • User-Friendly Electronic Pipetting: Whether you are new to the lab or an expert, the 8-channel Pipette has something to offer every lab professional. It's as easy to start with as a mechanical pipette and offers incredible options for even the most advanced user. You can utilize the build-in plate tracker to increase reliability in microwell plate work.
  • Ready to use: The 8-channel Picus™ Electronic Pipette comes ready to use and partially charged (however, we recommend fully charging it before its first use). For more information on charging, storage, and specifications, consult with Sartorius personnel and additional material available on our site.
  • Reliable and repeatable pipetting results
  • A wide selection of pipetting modes
  • Customizable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
TRUSTED_SUSTAINABILITY

Specifications

Compatible Tips Sartorius Optifit Tips, SafetySpace Tips, Flexibulk Tips
Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Gray
Disposable Reusable
For Use With Optifit Tip (10μL), SafetySpace Filter Tip
Grip Type None
Material Polycarbonate ABS
Number of Channels 8
Programmability Yes
Sterility Non-sterile
Type Electronic Pipette
Volume (Metric) 300 μL
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.