missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ 0.1% Formic Acid (v/v) in Acetonitrile, LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
Brand: Thermo Scientific™ 85174
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Thermo Scientific™ 0.1% Formic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.
This 0.1% Formic Acid in Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography- and spectrometry-grade 0.1% FA in acetonitrile is 0.2–micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection. (CAS: 64-18-6 )Highlights:
Validated – tested more than 20 ways to ensure maximal LC/MS sensitivity
Purified – processed, filtered and sealed to extend LC/MS column life
Convenient – packaged to eliminate variability and reduce handling
Specifica
LC-MS | |
1 L | |
Liquid | |
Pierce™ |
Formic Acid | |
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight. | |
Solvent Mixture | |
HPLC, Mass Spectrometry |
For Research Use Only. Not for use in diagnostic procedures.