missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile, LC-MS Grade

An LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

£207.00 - £704.00

Specifications

Grade LC-MS
Item Name Trifluoroacetic Acid (TFA)
Content And Storage Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Form Liquid
Reagent Type Solvent Mixture, Ion Pairing Reagent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
13444279
View Documents
Thermo Scientific™
85177
4 x 1 L N/A N/A N/A
13424279
View Documents
Thermo Scientific™
85176
1 L N/A N/A N/A
Description

Description

Thermo Scientific™ 0.1% Trifluoroacetic Acid in Acetonitrile is an LC/MS-grade preparation with high purity and low UV-absorptivity that is ideal for HPLC and mass spectrometry applications.

This 0.1% Trifluoroacetic Acid in Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance, providing the most sensitive detection across wavelengths and prolonging equipment life. The chromatography-and spectrometry-grade 0.1% TFA in acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • Validated

    – tested more than 20 ways to ensure maximal LC/MS sensitivity

  • Purified

    – processed, filtered and sealed to extend LC/MS column life

  • Convenient

    – packaged to eliminate variability and reduce handling

Specifications

Specifications

LC-MS
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Solvent Mixture, Ion Pairing Reagent
HPLC, Mass Spectrometry
Trifluoroacetic Acid (TFA)
Liquid
Pierce™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ 0.1% Trifluoroacetic Acid (v/v) in Acetonitrile, LC-MS Grade

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.