Learn More
Thermo Scientific™ Pierce™ 20X Modified Dulbecco's PBS Tween™ 20 Buffer
Prepare Na- and K-phosphate buffered saline (D-PBS) wash and blocking buffers for ELISA, Western and other immunoassays with this space-saving stock solution.
Brand: Thermo Scientific™ 28346
Code : NEW
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ Pierce 20X Modified Dulbecco PBS Tween-20 is a space-saving stock solution used to prepare physiological Na- and K-phosphate buffered saline (D-PBS) wash and blocking buffers for ELISA, Western and other immunoassays.
Thermo Scientific Pierce Concentrated Buffers are ready to use without having to reconstitute with ultrapure water and, in the case of Tris-Glycine Buffer, methanol. Buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation and other applications.Highlights:
Easy to use – no packets to open and no powder to dissolve
Increased accuracy – eliminates the possibility of powder remaining in a packet
Saves time – Concentration eliminates time spent waiting for powder to dissolve
Recommended for:
A wash buffer for ELISA, Western and other immunoassays as well as a blocking buffer for plate-based assays