Learn More
Thermo Scientific™ Pierce™ 660nm Protein Assay Reagent
Red-to-green (A660nm), ready-to-use, detergent- and reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
Brand: Thermo Scientific™ 22660
147.05 GBP valid until 2024-12-31
Use promo code "21989" to get your promotional price.
Code : PN
Additional Details : Weight : 1.36080kg
Description
Thermo Scientific™ Pierce 660nm Protein Assay is a quick, ready- to-use colorimetric method for total protein quantitation.
Additional Information and Online Ordering »
Although the Pierce 660nm Protein Assay produces a higher level of protein-to-protein variation (37%) than other assays, such as the BCA Protein Assay, the simpler single-reagent format and broader substance compatibility make the 660nm Assay more convenient for many routine applications. The Pierce 660nm Protein Assay can be performed in either a test tube or microplate format.
The assay is based on a proprietary dye-metal complex that binds to proteins in an acidic medium. Upon binding, the reddish dye-metal complex turns to green, resulting in an absorbance shift that is measurable at wavelengths near 660nm (645 to 670). The dye interacts mainly with basic residues in proteins, such as histidine, arginine and lysine and to a lesser extent tyrosine, tryptophan and phenylalanine.
Highlights:
Versatile – works with a greater range of detergents and reducing agents than other dye-based assays
Fast – single reagent with a simple mix-and-read protocol; no working reagent to prepare
Accurate – produces standard curves that are more linear than with the Bradford assay method
Flexible – assay may be performed in test tubes or microplates
Conserve samples – requires only 10μL for microplate or 100μL for the test tube procedures
Convenient – room temperature storage means no waiting for reagent equilibration before use
Includes:
Kit contains Pierce 660nm Assay Reagent and a set of seven pre-diluted albumin protein assay standards
Specifications
Store the assay reagent at room temperature. | |
Protein Assay Reagent | |
Pierce™ | |
Not Target-Specific | |
Solution-based Detection, Absorbance | |
750 mL |
Pierce 660nm Protein Assay Reagent | |
Colorimetric | |
Protein Quantitation Assay | |
500 Tube Assays or 5000 Microplate Assays | |
Spectrophotometer, Microplate Reader |
Safety and Handling
- Pierce(R) 660nm Protein Assay Reagent, 750 mL.
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol
For Research Use Only. Not for use in diagnostic procedures.