Learn More
Thermo Scientific™ Pierce™ 96-Well Polystyrene Plates, White Opaque
Bind antibodies with high capacity using these specially formulated polystyrene plates for ELISA and other immunoassays.
Brand: Thermo Scientific™ 15042
112.00 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Pierce 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics.
Thermo Scientific Pierce White Opaque 96-Well Microplates are made of specially formulated polystyrene and treated to produce high-capacity antibody-binding characteristics. These microplates have a corner notch for use in standard plate readers.
Features of White Opaque 96-Well Microplates:
- White opaque plates for fluorescent or chemiluminescent assays
- Treated to provide protein coating capacity
- High antibody-binding surface improves the overall performance in immunoassay applications
- Plates are certified for performance, having consistently low coefficients of variation among wells
Plate Properties:
- Antibody binding capacity: High, 400ng IgG/cm2 (1 cm2 = 100 μL volume)
- Binding interaction: hydrophobic-ionic
- Well-to-well coefficient of variation: CV <10%
- Well shape (bottom): Flat
- Maximum well volume: 360 μL
Related Products
- Pierce™ 96-Well Polystyrene Plates, Corner Notch (Cat. No. 15041)
Specifications
Adsorption, Hydrophilic | |
Colorimetric | |
Pierce | |
Well Plate | |
360 μL | |
Non-sterile | |
Flat Bottom (F-well) |
Pierce 96-Well Polystyrene Plates, White Opaque | |
25 Plate | |
White | |
Polystyrene | |
96 | |
Polystyrene | |
None |
For Research Use Only. Not for use in diagnostic procedures.