missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 1-Step™ ABTS Substrate Solution

Product Code. 10269423 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 10269423

Brand: Thermo Scientific™ 37615

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Ready-to-use, water-soluble peroxidase substrate tablets that yield a measurable green end-product for use in ELISA methods.

Thermo Scientific™ Pierce ABTS Substrates are tablets and a ready-to-use solution of a water-soluble peroxidase substrate that yields a measurable green end product for use in ELISA methods.

ABTS (2,2'-Azinobis [3-ethylbenzothiazoline-6-sulfonic acid]-diammonium salt) is a water-soluble HRP substrate that yields a green end product upon reaction with peroxidase. The green product has two major absorbance peaks, 410nm and 650nm. ABTS is less sensitive than OPD and TMB in ELISA applications. It is less readily oxidized, and its color development is slower (approximately 20 minutes). This may be advantageous if unacceptable background results from the use of the OPD or TMB substrates due to higher sensitivities.

Highlights:

  • Excellent choice when maximum sensitivities are not required
  • Less readily oxidized and color development is slower (approximately 20 minutes) than OPD and TMB
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce 1-Step ABTS Solution
Product Type ELISA Substrate
Quantity 250 mL
Detection Method Colorimetric
Product Line 1-Step
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
Content And Storage Store at 4°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ABTS Substrate Solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-iminodiethanol,sodium azide

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.