missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Acetonitrile (ACN), LC-MS Grade

An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Brand:  Thermo Scientific™ TS-51101


 View more versions of this product

Product Code. 11581094

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:
  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
  • Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
  • Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials

Product Details:

  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780 g/mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LC-MS
1 L
Liquid
Pierce™
75-05-8
Acetonitrile (ACN)
Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials.
Solvent (Pure)
HPLC, Mass Spectrometry
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.