missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Activated Peroxidase Antibody Labeling Kit

Glutaraldehyde-activated horseradish peroxidase (HRP) enzyme and kit to label primary antibodies and purify the resulting antibody-HRP conjugates.

Brand:  Thermo Scientific™ 31497

Product Code. 10035883

  • £674.00 / 1 set
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

Availability may vary by country

This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Activated HRP and Antibody Labeling Kit contains horseradish peroxidase enzyme that has been glutaraldehyde-activated to enable labeling and preparation of antibody conjugates.

This chemistry can be used to couple HRP to primary amines on another protein such as an antibody. The Thermo Scientific Activated Peroxidase Kit is designed to purify HRP-antibody conjugates by using Immobilized Protein A/G to separate unreacted enzyme from the antibody conjugate. Using Activated HRP produces an HRP-antibody conjugate in which both the enzyme activity and the antigen-binding activity are preserved.

Highlights:

  • Reacts with primary amines to form a covalent amide bond
  • No reduction step is necessary to secure the linkage
  • Can be stored for at least 12 months at -20'C
  • One mg produces about 0.5mL of conjugate with a working dilution of 1:1,000 when coupled to a high titer antibody
  • Enzyme activity is > 200 units/mg

Includes:

5mg Activated Peroxidase, 50mL Conjugation Buffer, 250mg Lysine, 0.5mL Immobilized Protein A/G Column, 200mL Gentle Ag/Ab Binding Buffer, 200mL Gentle Ag/Ab Elution Buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Antibody Labeling Kit
5 conjugations, each with 0.3 mg antibody (IgG)
300 μg
HRP (Horseradish Peroxidase)
5 reactions
Sufficient For: 5 conjugations, each with 0.3 mg antibody (IgG)
  • Activated Peroxidase, 5 mg
  • Conjugation Buffer, 50 mL
  • Lysine, 250 mg
  • Protein A/G Column, 0.5 mL
  • Gentle Ag/Ab Binding Buffer, 200 mL
  • Gentle Ag/Ab Elution Buffer, 200 mL

    Store activated peroxidase at -20°C. Store other components at 4°C.
  • EZ-Link™
    IgG
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.