missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Albumin Serum Depletion Kits

Optimize serum sample prep by depleting unwanted albumin and IgG serum proteins for protein-profiling methods using quality depletion kits.

Brand:  Thermo Scientific™ 89875

431.68 GBP valid until 2024-12-31
Use promo code "22006" to get your promotional price.



 View more versions of this product

Product Code. 10579853

  • £456.00 / 1 set
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Concentrations of albumin and IgG can make up more than 70% of total serum proteins and often mask lower quantity proteins during protein-profiling experiments. Optimize your serum sample prep by removing albumin or IgG quickly and conveniently. By using our depletion kits to remove excess high-abundance serum proteins, less abundant proteins of interest can be easily detected by 2D electrophoresis and other protein profiling methods such as mass spectrometry.

Pierce Albumin/IgG Removal Kit

Optimized—microcentrifuge spin cup method is scaled for treating 10 μL human serum samples for 2D electrophoresis
Efficient—removes nearly all HSA and IgG, enabling low-abundance proteins to be detected in 2D gels
Fast—processes samples in less than 40 minutes
Scalable—use different amounts of affinity resin to match sample volume and concentration

The Thermo Scientific Pierce Albumin/IgG Removal Kit uses a classical mixture of Cibacron Blue Dye and Protein A resin for removal of albumin and IgG. The dye binds to albumin with good specificity, and Protein A binds to many species and subclasses of IgG. The Pierce Albumin/IgG Removal Kit is economical and can be used to treat samples from other species besides human (e.g., monkey, swine, rabbit). The kit is also scalable and sufficient to process 25 samples, each containing 600 μg of total serum protein (about 10 μL of serum).

Pierce Albumin Depletion Kit

Complete—includes optimized buffer and microcentrifuge spin columns to remove albumin quickly and conveniently from 10- to 50-μL samples
Adaptable—easy-to-dispense slurry improves performance, eases processing of multiple samples, and can be adapted to larger or smaller columns or 96-well filter plates
Compatible—removing excess albumin facilitates MS analysis or electrophoresis gel analysis of low-abundance serum proteins

The Thermo Scientific Pierce Albumin Depletion Kit resin is supplied as a 50% slurry. To prepare the depletion columns, 200 μL of slurry is dispensed into the supplied microcentrifuge spin columns to obtain 100 μL of settled beads to process 10 to 50 μL of serum sample in a single reaction. The kit has been optimized to bind human, porcine, and sheep albumin from serum samples. With a modification to the protocol, albumin from bovine, calf, goat, and rat can be removed with this method. The kit is not effective for removal of mouse albumin. Processed samples are ready for immediate downstream analysis by electrophoresis or MS applications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Albumin and IgG Removal Kit
Albumin/Antibody Removal Kit
Upon receipt, store at 4°C. Do not freeze.
Product shipped with an ice pack.
Protein
Spin Column Kit
25 Reactions
Microcentrifuge, mass spectrometer, 2D gel electrophoresis
Pierce™
Cibacron™ Blue Dye, Protein A
Plasma, Serum
2D Gel Electrophoresis, Mass Spectrometry
Sample preparation
Serum Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Albumin Serum Depletion Kits

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Sodium chloride

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.