Learn More
Thermo Scientific™ Ammonium Persulfate
Catalyze acrylamide and bisacrylamide polymerization when preparing polyacrylamide gels for electrophoresis with APS oxidizing agent.
Brand: Thermo Scientific™ 17874
53.24 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : CAS Number : 7727-54-0 Weight : 0.10000kg Transport : UN number : 1444 Chem class : 5.1 Pack group : III
Description
Thermo Scientific™ Pierce Ammonium Persulfate (APS) is an oxidizing agent that is used with TEMED to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.
Ammonium persulfate (APS) is an oxidizing agent that is often used with tetramethylethylenediamine (TEMED, Part No. 17919) to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.Highlights:
- Name: Ammonium persulfate (APS)
- Formula: (NH4)2S2O8
- CAS number: 7727-54-0
- Molecular weight: 228.2
- Purity: ≥98.0%
- Insolubles: ≤0.005%
Compatible with:
TEMED, Part No. 17919
Specifications
Store below 23°C. | |
Ammonium Persulfate | |
Powder |
Safety and Handling
- Ammonium persulfate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : diammonium peroxodisulphate
For Research Use Only. Not for use in diagnostic procedures.