missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Miser™ Antibody Extender Solution NC

Product Code. 10500634 Shop All Thermo Scientific Products
Click to view available options
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 10500634

Brand: Thermo Scientific™ 32110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enable Western blot detection with much less primary antibody by treating nitrocellulose membrane with this reagent.

Perform Western blot detection with much less primary antibody using Thermo Scientific™ Pierce Antibody Extender Solution NC, a post-transfer nitrocellulose membrane-treatment reagent.

By treating nitrocellulose membranes for a few minutes in this reagent immediately after transfer and before probing, you can use much less (more dilute) primary antibody solutions while obtaining the same level of detection. This conserves use of expensive and rare antibodies, enabling more experiments to be performed with the same amount of antibody. The reagent is so effective that our scientists have observed antibody cost reductions of 10-, 25- and even 100-times (depending on the antigen and primary antibody directed against that antigen).

Highlights:

  • Achieve equivalent signal – use at least three times less primary antibody than normal and retain comparable or better signal detectability

  • Any detection mode or enzyme – the method works with both colorimetric and chemiluminescent detection modes and with horseradish peroxidase (HRP) and alkaline phosphatase (AP) systems

  • Easy-to-perform, 10-minute protocol – invest a few minutes and save money on your primary antibody; our scientists have reduced their use of expensive primary antibodies by up to 100-fold with this product

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Extender Solution
Content And Storage Store at room temperature.
Form Liquid
Membrane Nitrocellulose Membrane
Product Line Miser
For Use With (Application) ELISA, Western Blot
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.