missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Avidin

Perform Western blot, ELISA or other biotin-based detection methods using these purified and HRP-, AP- or FITC-labeled avidin molecules.

Brand:  Thermo Scientific™ 21121

119.70 GBP valid until 2025-12-31
Use promo code "24369" to get your promotional price.


 View more versions of this product

Product Code. 11836814

  • £133.00 / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Avidin and Conjugates include purified avidin protein from hen egg whites, plus avidin-HRP, avidin-AP and other conjugated forms for use in biotin-based detection methods.

Avidin from chicken egg whites is a glycoprotein that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods. Pure avidin is a key component in certain immunohistochemistry (IHC) methods. Alkaline phosphatase (AP) and horseradish peroxidase (HRP) conjugates of avidin are useful for Western blotting or ELISA detection with appropriate enzyme substrates.

Highlights:

  • Native avidin – glycoprotein from chicken egg whites, 67kDa, pI equals 10

  • Soluble – glycosylation and isoelectric point renders avidin more soluble than streptavidin

  • Affordable – significantly less expensive than recombinant streptavidin

  • ABC staining – avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)

  • Pure protein and conjugates – choose pure, unconjugated avidin for use in ABC staining or peroxidase or alkaline phosphatase forms for membrane and plate-based assays

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Unconjugated
Store at -20°C. Shipped at ambient temperature.
Pierce™
Biotin
10 mg
Solid
Avidin (Unconjugated)
Unlabeled
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL404863 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number21121
Lot NumberZH392891 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number21121
Lot NumberZA388591 Certificate TypeCertificate of Analysis Date27/01/2024 Catalog Number21121
Lot NumberYL362969 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number21121
Lot NumberYL362969 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number21121
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    40

    Hours

    14

    Minutes

    12

    Seconds

    2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Avidin > Avidin; 10mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.