Learn More
Thermo Scientific™ Pierce™ Background Eliminator Kit
Rapidly remove haze and artifacts from X-ray film to correct overexposed blots, whether from chemiluminescent Western blot or DNA or RNA autoradiography.
Brand: Thermo Scientific™ 21065
Code : PN
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Pierce Background Eliminator provides fast, easy removal of haze and artifacts on developed X-ray film to correct for overexposure of blots, whether from chemiluminescent Western blotting or DNA or RNA autoradiography.
This two-reagent kit is simple to use. Mix the two reagents (1:1 ratio) and immerse a developed piece of film (recent or old) in the working solution. Watch for 1 to 5 minutes until the desired image appears, then stop the reaction by rinsing the film in water. Pierce Background Eliminator for Film evenly reverses development-signal over the entire film, preserving relative densitometry values so that analysis and publication of results is still valid and possible.Highlights:
For overexposed X-ray film – reverses development of silver-based autoradiography film removing silver emulsion from the film surface
Recovers and preserved data – evenly reduces developed signal over the entire film so relative intensity (densitometry) of bands is not altered
Optimizes for signal-to-noise – allows readjustment of overdeveloped film to obtain the best image with respect to background haze, speckles and oversaturated bands that need to be resolved
Saves time and resources – eliminates the need to repeat experiments or re-expose a new piece of film to obtain an optimal image
Recommended for:
Remove background from any application that uses X-ray film exposures including: Western, Northern and Southern blots that use Thermo Scientific SuperSignal Substrates and the Pierce Chemiluminescent Hybridization and Detection Kit; In-gel detection systems; Gel-shift assays; Ribonuclease protection assays (RPA)
Specifications
Background Eliminator Kit | |
Western Blot | |
Sufficient For: 2.5L of 1X solution to treat at least 10 films (13 x 18 cm ea.) • Reagent A, 100 mL • Reagent B, 100 mL Store at room temperature |
Pierce | |
All species |
For Research Use Only. Not for use in diagnostic procedures.