missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotin

Purified biotin and biotinylated BSA, HRP, AP or FITC for use as controls or to amplify signal in IHC via the avidin-biotin complex (ABC).

Brand:  Thermo Scientific™ 29129

76.95 GBP valid until 2025-12-31
Use promo code "24369" to get your promotional price.


Product Code. 10588733

  • £85.25 / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.

Biotinylated HRP and biotinylated AP are most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method. Biotinylated Fluorescein (FITC) cannot be used to polymerize and amplify the signal to the same degree as biotinylated enzymes because each fluor molecule is tagged with only one biotin molecule. However, the fluorescent variant of traditional ABC staining is possible with this unique molecule.

Highlights:

  • Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining

  • Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)

  • Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Biotin
1 g
Carboxylic Acid
Powder
Biotin (Unconjugated)
Pierce Biotin
Store in a cool, dry area, protected from direct sunlight.
DMSO (Dimethylsulfoxide), Water, DMF (Dimethylformamide)
Pierce™
Biotin & Analog
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ394470 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number29129
Lot NumberZH388845 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number29129
Lot NumberZE388844 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog Number29129
Lot NumberZB384568 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number29129
Lot NumberYK380459 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number29129
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Biotin > Biotin; 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.