missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Biotin-Fluorescein Conjugate

Product Code. 10752905 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 10752905

Brand: Thermo Scientific™ 22030

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purified biotin and biotinylated BSA, HRP, AP or FITC for use as controls or to amplify signal in IHC via the avidin-biotin complex (ABC).

Thermo Scientific™ Pierce Biotinylated Proteins include biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and fluorescein (B-FITC) for use as controls or signal amplification in IHC via avidin-biotin complex (ABC) techniques.

Biotinylated HRP and biotinylated AP are most commonly used in immunohistochemistry (IHC) to amplify the signal of biotinylated primary antibodies using the ABC staining method. Biotinylated Fluorescein (FITC) cannot be used to polymerize and amplify the signal to the same degree as biotinylated enzymes because each fluor molecule is tagged with only one biotin molecule. However, the fluorescent variant of traditional ABC staining is possible with this unique molecule.

Highlights:

  • Biotinylated enzymes – biotin-labeled horseradish peroxidase (B-HRP), alkaline phosphatase (B-AP) and beta-galactosidase (B-bGal) for use in avidin-biotin complex (ABC) staining

  • Biotinylated FITC – amplify the fluorescein signal of biotinylated primary antibodies by using avidin or streptavidin plus biotinylated fluorescein (B-FITC)

  • Biotinylated controls – use biotinylated bovine serum albumin (B-BSA) or any one of these molecules as positive controls in biotin quantitation methods

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Classic Dyes, Biotin and Analogs
Description Pierce Biotin-Fluorescein Conjugate
Quantity 5 mg
Content And Storage Store at 4°C, protected from moisture. Shipped at room temperature.
Excitation/Emission 494/518
Solubility DMF (Dimethylformamide)
Form Solid
Product Line Pierce
Product Type Biotin Conjugate (Fluorescein)
compliance-icons
Product Identifier
  • Biotin-Fluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Biotin-Fluorescein

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.