Learn More
Thermo Scientific™ BM(PEG)3 (1,11-bismaleimido-triethyleneglycol)
Crosslink sulfhydryls to sulfhydryls (cysteine residues) with this long, pegylated, water-soluble crosslinker composed of maleimide groups and 3-unit (17.8A) PEG spacer arm.
Brand: Thermo Scientific™ 22337
Code : PN
Additional Details : Weight : 2.72160kg
Description
Thermo Scientific™ Pierce BM(PEG)3 is a pegylated crosslinker having the form maleimide-PEG-maleimide for conjugation between sulfhydryl groups (e.g., cysteines) via a three-unit polyethylene glycol (PEG) spacer arm (17.8 Ås).
Highlights:
- Long sulfhydryl-reactive, homobifunctional crosslinker
- Water-soluble
- Polyethylene glycol (PEG) spacer provides increased water solubility, reducing the potential for crosslinker-caused precipitation
- Ideal for small molecule or peptide conjugations
- Reactive groups: maleimide (homobifunctional)
- Reactive toward: sulfhydryl groups
Specifications
Sulfhydryl-Sulfhydryl | |
BM(PEG)3 | |
Yes | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Maleimide | |
Solid | |
Water | |
Yes |
No | |
352.34 | |
17.8 Å | |
No | |
Pierce™ | |
Homobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.