missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ CN/DAB Substrate Kit

Combines chloronaphthol- and diaminobenzidine-based HRP detection for Western blot and tissue staining methods.

Brand:  Thermo Scientific™ 34000


Product Code. 10322284

  • £149.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce CN/DAB Substrate Kit includes substrate and peroxide solutions for combined chloronaphthol- and diaminobenzidine-based detection of horseradish peroxidase (HRP) activity in blotting and tissue staining methods.

This kit combines two popular chromogenic peroxidase substrate compounds (4-CN and DAB) in a single, stable, 10X stock solution. This CN/DAB solution produces an intense dark black precipitate at sites of bound HRP-conjugated antibodies on probed blots and tissue samples. The Pierce CN/DAB Substrate Kit can be used in Western immunoblotting techniques as well as in dot blotting assays. The kit includes the 10X solution of CN/DAB and accompanying 1X Stable Peroxide Substrate Buffer.

Highlights:

  • HRP substrate – for detection of horseradish peroxidase activity on solid media via the combined action of CN (4-chloro-1-naphthol) and DAB (3,3'-diaminobenzidine tetrahydrochloride)

  • Chromogenic – no special equipment needed for visualization; yields intense black bands that are easy to photograph

  • Easy-to-use kit – substrate stock solution and stable peroxide substrate buffer mean that there is no dry powders to measure and dissolve

  • Stable – store refrigerated at 4°C for at least one year

Includes:

10X solution of CN/DAB and the Pierce Stable Peroxide Substrate Buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Substrate
Liquid
275 mL
Horseradish Peroxidase
25 Uses at 10 mL Each
Colorimetric
Pierce™
HRP
Horseradish Peroxidase
Sufficient For: 25 uses at 10 mL each
• CN/DAB (10X), 25 mL
• Stable Peroxide Buffer, 250 mL

Store at 4°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CN/DAB Substrate Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.