Learn More
Thermo Scientific™ Pierce™ Color Silver Stain Kit
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Color Silver Stain Kit enables sensitive detection of proteins electrophoresed in 1D and 2D polyacrylamide gels using a metallic silver technique which produces color variations that facilitate differentiation of protein bands and spots.
Highlights:
Customizable – kit is a reagent set that allows customized formulation of working reagents and staining protocols for particular gel formats and silver-staining applications
Sensitive – detects many proteins at 0.1ng per square millimeter in standard-thickness gels
Broad protein detectability – stains a wider range of protein types than monochromatic silver stains (proteins that do not bind silver are revealed as yellow bands or spots)
- Color differentiation – gel-separated proteins and polypeptide-containing macromolecules stain in five basic colors: black, blue, brown, red and yellow
Profiling resolution – multicolored detection of DNA, lipids and polysaccharides in addition to typical proteins provides additional landmarks to aid in 2D gel profiling
Includes:
Four reagent solutions (500mL each): silver, reducer base, reducer aldehyde and stabilizer base
Specifications
Specifications
| Content And Storage | Kit contains sufficient reagents to stain 25 (18 cm2) or 40 (130 cm2) gels • 500 mL Silver Reagent • 500 mL Reducer Aldehyde Reagent • 500 mL Reducer Base Reagent • 500 mL Stabilizer Base Reagent |
| Detection Location | In-Gel Detection |
| Detection Method | Colorimetric |
| Label or Dye | Silver |
| Product Type | Stain Kit |
| Product Line | Pierce |
| Quantity | 2 L |
| Target Molecule | Protein |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.