missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Compat-Able™ BCA Protein Assay Kit

Product Code. 10688484 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10688484

Brand: Thermo Scientific™ 23229

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precipitation reagents to eliminate interfering substances from protein assay samples to make them compatible with BCA and Coomassie-Bradford assays.

Thermo Scientific™ Compat-Able Protein Assay Preparation Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.

The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps. The Compat-Able Preparation Set is available individually or conveniently bundled with either the Thermo Scientific Pierce BCA Protein Assay Kit or the Thermo Scientific Pierce Coomassie Plus Protein Assay Kit.

Highlights:

  • Eliminate incompatibilities – removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer

  • Simple and fast – prepare samples in less than 10 minutes with four simple steps

  • Convenient – reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results

  • Scalable – microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs

  • Validated – tested and offered together with BCA and Coomassie Plus Protein Assay Kits; protocol integrates control and normalization procedures

Includes:

Set contains two bottled reagents (Reagent 1 and Reagent 2); Complete kits contain the Reagent Set plus components of the respective protein assay

TRUSTED_SUSTAINABILITY

Spezifikation

Content And Storage Sufficient For: Treating up to 500 protein assay samples and performing 250 tube assays or 2500 microplate assays
• Compat-Able Reagent 1, 250 mL
• Compat-Able Reagent 2, 250 mL
• BCA Reagent A, 500 mL
• BCA Reagent B, 25 mL
• Albumin Standard Ampules, 2 mg/mL, 10 x 1 mL
Description Compat-Able BCA Protein Assay Kit
Assay BCA Assay
Detection Method Colorimetric
Product Line Compat-Able
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 250 Tube Assays or 2500 Microplate Assays
For Use With (Application) Absorbance, Solution-based Detection, Incompatibility Elimination
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 1 L
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt