Learn More
Thermo Scientific™ Pierce™ Bradford Protein Assay Kit
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Measure protein concentration using a stable formulation of the original Bradford reagent described in the literature with Thermo Scientific™ Pierce Coomassie Protein Assay Kit.
Highlights:
Bradford reagent – stable, ready-to-use kit of the classical Bradford assay reagent
Colorimetric – measure with a standard spectrophotometer or plate reader (595nm)
Easy to use – single reagent; no working reagent preparation required
Fast – almost immediate color development; add, mix and read results
Broad range – detects protein concentration in the range 1 to 1500μg/mL
Flexible – microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges
Includes:
Coomassie Protein Assay Reagent and 10 ampules of Albumin Standard
Specifica
Specifica
| Content And Storage | Sufficient For:
|
| Assay | Bradford Assay |
| Detection Method | Colorimetric |
| Product Line | Pierce |
| Product Type | Protein Quantitation Assay |
| Specificity | Not Target-Specific |
| Sufficient For | 630 Tube Assays or 3800 Microplate Assays |
| For Use With (Application) | Solution-based Detection, Absorbance |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Quantity | 950 mL |
Sicurezza e movimentazione
- Pierce Coomassie (Bradford) Protein Assay Kit
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 2
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H371-May cause damage to organs.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305-IF IN EYES:.
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : phosphoric acid, methanol
For Research Use Only. Not for use in diagnostic procedures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.