Learn More
Thermo Scientific™ Pierce™ Bradford Plus Protein Assay Reagent
Brown-to-purple (A595nm), ready-to-use, fast reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.
Brand: Thermo Scientific™ 23238
161.10 GBP valid until 2024-12-31
Use promo code "21989" to get your promotional price.
Code : PN
Additional Details : CAS Number : 7664-38-2 Weight : 1.47420kg Transport : UN number : 3264 Chem class : 8 Pack group : II
Description
Thermo Scientific™ Pierce Coomassie Plus Protein Assay is a high-performance Bradford assay reagent — one of the fastest, simplest and most popular protein quantitation methods available.
The Pierce Coomassie Plus Assay Reagent is a single, ready-to-use solution for measuring protein concentration. Simply add the reagent to equal volumes of samples and standards, mix, then measure the absorbance at 595nm. The assay costs only pennies per sample and can be performed in either test tube or microplate format. The Pierce Coomassie Plus Assay Reagent provides increased linearity of response and only half the protein-to-protein variability of other commercial Bradford assay formulations.Highlights:
Colorimetric – measure with a standard spectrophotometer or plate reader (595nm)
Easy to use – single reagent; no working reagent preparation required
Fast – almost immediate color development; add, mix, and read results
Broad range – detects protein concentration in the range 1 to 1500μg/mL
Better – improved linearity and response-uniformity compared to traditional Bradford formulations
Flexible – microplate and cuvet protocols provided with the instructions and adaptable to several target working ranges
Specifications
Store at 4°C. | |
Bradford Assay | |
Pierce™ | |
Not Target-Specific | |
Spectrophotometer, Microplate Reader |
Pierce Bradford Plus Protein Assay Reagent | |
Colorimetric | |
Protein Quantitation Assay | |
Solution-based Detection, Absorbance | |
300 mL |
Safety and Handling
- Coomassie PlusTM Protein Assay Reagent
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H371-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P305-IF IN EYES:.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : phosphoric acid, methanol
For Research Use Only. Not for use in diagnostic procedures.