missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Crossflow Filtration Cassettes, 3–100 kDa MWCO

Product Code. p-200006950 Shop All Thermo Scientific Products
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 18091673

Brand: Thermo Scientific™ A57930

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Pierce Crossflow Cassettes are membrane-based devices designed for concentrating and diafiltration of sample solutions such as proteins, lipids, viruses, and cell culture supernatants with volumes from 0.50–3 L. Suited for manufacturing and pilot-scale tangential flow filtration processes.

Pierce Crossflow Cassettes are designed for easy setup, flexibility, and robust performance. They can process sample solutions ranging from 0.5 to 3 liters in volume and operate via a peristaltic pump pressure-driven process where the sample solution containing solutes are pushed tangentially across the surface of a specialized Polyethersulfone (PES) membrane with low protein binding characteristics. The membrane surface per device is 200 cm2 and is offered in five different molecular-weight cutoffs (MWCOs) of 3, 10, 30, 50, and 100 kDa, providing a range of options to suit various needs.

Thermo Scientific Pierce Crossflow Filtration Cassettes are membrane-based devices designed for small-volume manufacturing and pilot-scale tangential flow filtration (TFF) of biological samples. They feature easy setup, optimal flexibility, and robust performance. These cassettes can process sample solution volumes of 0.5–3 liters and operate via a peristaltic pump pressure-driven process where the sample solution is pushed tangentially across the surface of a specialized polyethersulfone (PES) membrane.

Features of Pierce Crossflow Filtration Cassettes include:
• Designed for small-volume manufacturing and pilot-scale TFF applications
• Reusable without membrane replacement
• Fully assembled with easy setup requiring no holder
• High flow rate and sample recovery
• Low cost of operation

Pierce Crossflow Filtration Cassettes contain a PES membrane with low protein binding characteristics and have a specialized channel design that ensures high flux without optimization. The membrane has a surface area of 200 cm2 and the option of five different molecular-weight cutoffs (MWCOs): 3, 10, 30, 50, and 100 kDa. The membrane has small pores, preventing the passage of larger molecules like proteins that are retained in the flow feed path while smaller molecules like salts pass through, causing a decrease in solution volume and an increase in macromolecule concentration.

Pierce Crossflow Filtration Cassettes are easily cleaned and reusable. Each cassette can concentrate up to 1 liter of sample by a factor of >50 in 30 minutes or less. Two cassettes can be connected and run in parallel using a Y connector to scale and speed up the process. In this way, up to 6L of sample can be processed.

Each cassette includes all necessary accessories required for running the cassette with a laboratory peristaltic pump and a size 16 pump head. To make the buffer exchange process more convenient, a sealed diafiltration reservoir accessory can be used to help speed up the process.

Pierce Crossflow Filtration Cassettes are designed for small-volume manufacturing and pilot-scale TFF applications in diverse biological and biopharmaceutical processes, such as concentration and diafiltration of proteins, lipids, viruses, colloids, and cell culture supernatants.  The cassettes deliver efficient performance due to their high crossflow speed and low pump speed requirement.

TRUSTED_SUSTAINABILITY

Tekniske data

Filter Type Membrane Filter
Format Cassette
For Use With (Equipment) Labscale TFF system
Material Polypropylene
Material (Membrane) PES
MWCO 3 kDa
Pore Size 1.2 nm
Sterility Non-sterile
Fit for Purpose Certified Quality
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.