missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Cypridina Luciferase Flash Assay Kit

Product Code. p-4532159 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Conditionnement:
Each
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Code produit. 12385613

Marque: Thermo Scientific™ 16168

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Assay regulatory element transcriptional activity in mammalian cell culture media and whole cell lysate with this highly sensitive flash-based luciferase kit.

The Thermo Scientific™ Pierce Cypridina Luciferase Flash Assay Kit provides researchers with a highly sensitive assay for transcriptional activity of regulatory elements in mammalian cell culture media and whole cell lysate.

This Flash Assay Kit contains reagents for measuring the activity of Cypridina luciferase in mammalian cell culture media and lysates. When used with Thermo Scientific Cypridina Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for secreted or intracellular detection of promoter or pathway activity. Cypridina luciferase is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. The signal produced by Cypridina is considerably greater than signal from either firefly or Renilla luciferases assayed under similar conditions.

Highlights:

  • Sensitive – high sensitivity allows utilization of smaller numbers of cells
  • Cost effective – sensitive assays result in decreased reagent consumption
  • Secreted luciferase – allows real-time assays and kinetic studies without destroying cells
  • Ideal for co-transfection – uses a different substrate than Renilla, Gaussia, or firefly luciferases
  • Compatible – assay reagents compatible with other Cypridina or Vargula luciferases
  • Time-saving – assays using secreted luciferases require minimal sample handling
  • Automation-friendly – amenable to high throughput screens
  • Convenient – contains a universal cell lysis buffer and optimized flash assay reagent
  • Safe – allows one to perform non-radioactive assays

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Cypridina luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers.

Recommended for:

Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Secretory pathway / protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies

TRUSTED_SUSTAINABILITY

Spécification

Type Assay Kit
Target Luciferase, Cypridina Luciferase
Quantity 100 Reaction Kit
Content And Storage • Cypridina Flash Assay Buffer, 5 mL, store at 4°C
• Vargulin (1X), 5 μL, store at -20°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature

Upon receipt store kit at -20°C or store individual components as indicated above.
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Vargulin (Cypridina Luciferin)
Afficher plus Afficher moins

For Research Use Only. Not for use in diagnostic procedures.

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis