Learn More
Thermo Scientific™ Pierce™ DAB Substrate Kit
Chromogenic horseradish peroxidase (HRP) substrate for Western blot and tissue staining methods.
Brand: Thermo Scientific™ 34002
117.80 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ Pierce DAB and the DAB Substrate Kit enable chromogenic detection of horseradish peroxidase (HRP) activity based the action of 3,3'-diaminobenzidine (DAB) in blotting and tissue staining methods.
DAB (3,3'-diaminobenzidine tetrahydrochloride; MW = 214.1) reacts with HRP in the presence of peroxide to yield an insoluble brown-colored product at locations where peroxidase-conjugated antibodies are bound to samples. The brown precipitate is insoluble in alcohol and other organic solvents, making it an excellent substrate for immunohistochemical staining that requires the use of traditional counterstains and mounting media. The Pierce DAB Substrate Kit includes a 10X solution of DAB and Stable Peroxide Substrate Buffer. Crystalline DAB (powder) is also available.Highlights:
HRP substrate – for detection of horseradish peroxidase activity on solid media, including nitrocellulose and PVDF membranes and fixed tissue samples
Chromogenic – no special equipment needed for visualization; the DAB reaction produces brown bands or spots at sites of reaction with HRP-conjugated antibodies or probes
Package options – choose powder for custom formulation with buffer and peroxide or a complete kit of preformulated solutions for immediate use
Specifications
Pierce DAB Substrate Kit | |
Colorimetric | |
Nitrocellulose, PVDF | |
275 mL | |
Horseradish Peroxidase | |
25 Uses at 10 mL Each |
Substrate | |
Liquid | |
Pierce™ | |
HRP | |
Horseradish Peroxidase | |
Sufficient For: 25 uses at 10 mL each • DAB (10X), 25 mL • Stable Peroxide Buffer, 250 mL Store kit at 4°C. |
Safety and Handling
- Pierce DAB Substrate Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P353-Rinse skin with water/shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol
For Research Use Only. Not for use in diagnostic procedures.