missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Dextran Desalting Columns (5K MWCO)

Ready-to-use, gel-filtration Thermo Scientific Pierce Dextran Desalting Columns can separate proteins and macromolecules from low-molecular weight buffer salts and reagents up to 5000 Daltons (5 kDa) in 5- or 10-mL volumes.

Brand:  Thermo Scientific™ 43233

 View more versions of this product

Product Code. 11839350

  • £124.00 / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Dextran Desalting Columns are ready-to-use, disposable, gel-filtration columns for separating proteins and other macromolecules from low molecular-weight buffer salts and reagents.

Applications
• Removing salts from protein solutions
• Eliminating phenol from nucleic acid preparations
• Separating excess crosslinker from conjugate preparations
• Removing excess derivatizing agents from modified proteins
• Withdrawing unreacted dye from fluorescent antibodies
• Eliminating free radiolabel from labeled proteins
• Buffer exchange

Gravity-flow gel filtration using dextran desalting columns
Gravity-flow gel filtration involves the chromatographic separation of molecules of different dimensions based on their relative abilities to penetrate a suitable stationary phase. A chromatographic matrix, usually consisting of very small, uncharged porous particles in an aqueous solution, is packed into a column and then used for separation via size exclusion. Different levels of separation can be achieved based on the pore size of the medium packed into the desalting column—in our case, dextran. With size exclusion filtration, large molecules are excluded from the internal pores of the gel and emerge from the column first. Smaller molecules can penetrate the pores, then progress through the column at a slower rate. These smaller molecules are subsequently flushed through the column with additional buffer volume. Our Dextran Desalting Columns are rigid for easy handling and provide excellent flow properties. Dextran is stable in water, salt solutions, organic solvents and alkaline or weakly acidic solutions. Dextran is also heat-stable and can be autoclaved dry or in solution at a neutral pH for 30 minutes at 120°C without affecting its chromatographic properties.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Desalting Column
Gravity Flow Column
Size-exclusion, Dextran Resin
Pierce™
5 Columns
Store at room temperature or 4°C.
Buffer Exchange, Protein
5.0 kDa
Single-use
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.