Learn More
Thermo Scientific™ Pierce™Dimethylsulfoxide (DMSO), Sequencing Grade
A sequencing-grade DMSO preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.
Brand:  Thermo Scientific™ TS-20688
Product Code. 10156644
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thermo Scientific™ Pierce™ Dimethylsulfoxide (DMSO) is a sequencing-grade preparation with high purity for use in traditional amino acid analyzers and other high-volume applications.
This sequencing-grade dimethylsulfoxide is >99.5% pure DMSO which is an excellent choice for protein and peptide applications that require the water-miscible solvent. The purity test ensures that preparation is essentially devoid of amines. This DMSO is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flammable
Room Temperature
950 mL
Pierce™
Dimethylsulfoxide (DMSO)
Liquid
99.5% Dimethylsulfoxide
LC/MS
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Pierce™Dimethylsulfoxide (DMSO), Sequencing Grade >
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.