missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DMP (dimethyl pimelimidate)

Product Code. p-4531007 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
This item is not returnable. View return policy

Product Code. 10097093

Brand: Thermo Scientific™ 21666

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Crosslink amines to amines with dimethyl pimelimidate (CAS 58537-94-3); composed of imidoester groups and a membrane-permeable, 7-atom spacer arm.

Thermo Scientific™ Pierce DMP is dimethyl pimelimidate, a membrane-permeable crosslinker that contains an amine-reactive imidoester group at each end of a 7-atom spacer arm.

Highlights:

  • Water-soluble
  • Rapid reaction with amines at alkaline pH values (pH 8-10)
  • Amidine bond retains net charge character of protein
  • Reversible at high pH values
  • Reactive groups: imidoester (homobifunctional)
  • Reactive toward: amino groups
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Description DMP
Molecular Weight (g/mol) 259.17
PEGylated No
Spacer Arm Length 9.2 Å
Content And Storage Upon receipt store desiccated at 4°C.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety Imidoester
Spacer Short (<10 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
compliance-icons
Product Identifier
  • DMP (dimethyl pimelimidate)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : dimethyl heptane-1,7-diimidate dihydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.