missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ DSG, No-Weigh™ Format

Product Code. 15859614 Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
10 x 1 mg
Unit Size:
50mg
Pack of 10
This item is not returnable. View return policy

Product Code. 15859614

Brand: Thermo Scientific™ A35392

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Thermo Scientific™ Pierce DSG is disuccinimidyl glutarate, a simple homobifunctional crosslinker based on the amine-reactive N-hydroxysuccinimide (NHS) ester group.

Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging helps prevent loss of reagent reactivity and contamination over time by minimizing repetitive opening and closing of vial. The format enables use of fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.

  • Reactive groups: NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO)
  • Membrane-permeable, allowing for intracellular crosslinking
  • Can increase crosslinking efficiency compared to DSS in some applications
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Amine
Cleavable No
Molecular Weight (g/mol) 326.26
PEGylated No
Spacer Arm Length 7.7 Å
Content And Storage Upon receipt store desiccated at 2 to 8°C.
Cell Permeability Yes
Shipping Condition Wet Ice
Product Line No-Weigh, Pierce
Labeling Method Chemical Labeling
Crosslinker Type Homobifunctional
Reactive Moiety NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 10 x 1 mg
Solubility DMF, DMSO
Format Standard, Single-use
Water Soluble No
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.