missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ DSP, No-Weigh™ Format

Crosslink amines to amines with dithiobis succinimidyl propionate, composed of NHS-ester groups and disulfide-containing, 12.0A spacer arm and cleavable with reducing agents.

£143.00 - £292.00

Specifications

Chemical Reactivity Amine-Amine
Cleavable By Thiols
Description DSP
Molecular Weight (g/mol) 404.42
PEGylated No
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15973902
View Documents
Thermo Scientific™
A35393
10 x 1 mg
£143.00
Pack of 10
In Stock
to see stock.
Add to basket
 
11821315
View Documents Promotion Details
Thermo Scientific™
22586
50 mg
£162.00
50mg
In Stock
to see stock.
Edge
Add to basket
 
10731945
View Documents
Thermo Scientific™
22585
1 g
£292.00
1g
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Thermo Scientific™ Pierce DSP Crosslinker (Lomant's Reagent) is an amine-reactive crosslinker with NHS-ester reactive ends and a cleavable disulfide bond in its spacer arm.

Highlights:

  • Cleavable by reducing agents
  • Membrane-permeable
  • Acylate in aqueous or organic media within two minutes at room temperature
  • Four- to five-hour half-life of ester at pH 7.0
  • Reactive groups: NHS ester (homobifunctional)
  • Reactive toward: amino groups
Specifications

Specifications

Amine-Amine
DSP
No
Upon receipt store desiccated at 4°C.
Chemical Labeling
NHS Ester
Powder
Standard, Single-use, Premium-grade
Crosslinker
By Thiols
404.42
12.0 Å
Yes
Homobifunctional
Medium (10 to 30 Å)
DMF, DMSO
No
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.