Learn More
Thermo Scientific™ Pierce™ ECL Western Blotting Substrate
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce ECL Western Blotting Substrate is a value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard enhanced chemilumescent (ECL) substrates for detection of horseradish peroxidase (HRP) enzyme activity, yet costs much less. Because the luminol and peroxide reagent-formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Simply switch to Pierce ECL and save a bundle.
Highlights:
ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes
About half the cost of other ECL Substrates – low overhead and a commitment to customer value enables Thermo Fisher Scientific to make this product available to you at a significantly lower price than other companies (these claims are based on the 2010 U.S. list prices)
No optimization required – switching to Pierce ECL substrate from other entry-level ECL substrates does not require optimization or protocol changes
A name you can can rely on – count on the strong quality assurance, technical support and reputation behind Thermo Scientific Pierce Protein Research Products
Includes:
Detection Reagent 1 and 2
Specifications
Specifications
| Product Type | Substrate |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| For Use With (Application) | Western Blot |
| Membrane Compatibility | Nitrocellulose, PVDF |
| Product Line | Pierce |
| Quantity | 250 mL |
| Substrate | Horseradish Peroxidase |
| Recommended Antibody Concentrations | 2°: 1:1-15K (0.07-1μg/mL), 1°: 1:1K (0.2- 10μg/mL) |
| Sensitivity | Low Picogram |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.