Learn More
Thermo Scientific™ Pierce™ EDC, No-Weigh™ Format
Immobilize or conjugate proteins using 1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide (CAS 25952-53-8), a water-soluble, carboxyl-to-amine crosslinker.
Brand: Thermo Scientific™ 22980
168.15 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce EDC is a water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation.
EDC is the abbreviation for 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, a crosslinker that has been used in diverse applications such as forming amide bonds in peptide synthesis, attaching haptens to carrier proteins to form immunogens, labeling nucleic acids through 5Æphosphate groups and creating amine-reactive NHS-esters of biomolecules. EDC reacts with a carboxyl to form an amine-reactive O-acylisourea intermediate. If this intermediate does not encounter an amine, it will hydrolyze and regenerate the carboxyl group. In the presence of N-hydroxysulfosuccinimide (Sulfo-NHS, Part No. 24510), EDC can be used to convert carboxyl groups to amine-reactive Sulfo-NHS esters.Highlights:
- Zero-length crosslinker
- Numerous chemical conjugates can be synthesized via available –COOH and –NH2 groups
- Amide bond formed provides a neutral linkage, which is ideal for preparing peptides and antigens
- Adding Sulfo-NHS (Part No. 24510) enhances the coupling reaction at physiological pH conditions
- Reactive groups: carbodiimide
- Reactive toward: carboxyl and amino groups

Specifications
Amine-Carboxyl | |
EDC | |
No | |
Upon receipt store desiccated at -20°C. | |
Wet Ice | |
Chemical Labeling | |
Carbodiimide | |
Powder | |
Water | |
Yes |
No | |
191.7 | |
0.0 Å | |
No | |
Pierce™ | |
Heterobifunctional | |
Short (<10 Å) | |
5 g | |
Standard, Single-use, Premium-grade | |
Crosslinker |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- 1-ETHYL-3-(3-DIMETHYLAMINOPROPYL)CARBODIIMIDE. HCl (EDC)
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Powering Proteomics
Advance your proteomics research with essential toolsPromo Code: 25977Expires: 2025-05-31
New Offer Save Up to 92%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.