missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Blocker Blocking Buffer

Increases signal-to-noise ratio and minimizes background

Brand:  Thermo Scientific™ N502

Product Code. 10627914

  • £277.00 / 1 L
Estimated Shipment: 28-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Reagent sold ready-to-use.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blocking Buffer
Liquid
Store at 2–8°C.
1 L
ELISA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL395567 Certificate TypeCertificate of Analysis Date21/12/2024 Catalog NumberN502
Lot NumberZJ392450 Certificate TypeCertificate of Analysis Date19/10/2024 Catalog NumberN502
Lot NumberZG388282 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberN502
Lot NumberZE386656 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog NumberN502
Lot NumberZC386568 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberN502
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ ELISA Blocker Blocking Buffer > ELISA Blocker; 1L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.